Assay Details
Gene Symbol: SH2D6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832332.1 | XP_028688165.1 | 1324 | 83 | ||
XM_028832333.1 | XP_028688166.1 | 1997 | 83 | |||
XM_028832334.1 | XP_028688167.1 | 1999 | 83 | |||
XM_028832335.1 | XP_028688168.1 | 2023 | 83 | |||
XM_028832336.1 | XP_028688169.1 | 1993 | 83 | |||
GenBank mRNA | JU342189.1 | - | 2 - 3 | 140 | 83 | |
JV712741.1 | - | 2 - 3 | 135 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
