Assay Details
Gene Symbol: SOX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002798399.3 | XP_002798445.1 | 999 | 96 | ||
GenBank mRNA | JU336056.1 | - | 3 - 4 | 906 | 96 | |
JV694275.1 | - | 3 - 4 | 977 | 96 |
Gene Symbol: SOX9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV721816.1 | - | 2 - 3 | 1060 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
