Assay Details
Gene Symbol: H6PD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001099298.4 | XP_001099298.2 | 1617 | 74 | ||
XM_015147201.2 | XP_015002687.1 | 1038 | 74 | |||
GenBank mRNA | JV076197.1 | - | 3 - 4 | 1019 | 74 | |
JV666219.1 | - | 3 - 4 | 991 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
