Assay Details
Gene Symbol: FAM228B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111814.2 | XP_014967300.2 | 616 | 135 | ||
GenBank mRNA | JU343160.1 | - | 5 - 6 | 560 | 135 | |
JV714358.1 | - | 5 - 6 | 503 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() |
