Assay Details
Gene Symbol: CDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122268.2 | XP_014977754.2 | 571 | 79 | ||
GenBank mRNA | JU332146.1 | - | 3 - 4 | 432 | 79 | |
JU478109.1 | - | 3 - 4 | 429 | 79 | ||
JV044671.1 | - | 3 - 4 | 369 | 79 | ||
JV564884.1 | - | 3 - 4 | 427 | 79 | ||
JV668548.1 | - | 3 - 4 | 453 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
