Assay Details
Gene Symbol: MIR563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032614.1 | 1 | 37 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | pleckstrin homology like domain family B member 2 | PHLDB2 | EGK_11279 | Anti-sense | Chr. 2 - 164202831 - 164443300 on NCBI Build Mmul_10 | |
Downstream | phosphatidylinositol specific phospholipase C X domain containing 2 | PLCXD2 | 46.2kb | Anti-sense | Chr. 2 - 164449238 - 164502342 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
