Assay Details
Gene Symbol: MIR572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032619.1 | 1 | 52 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kelch domain containing 4 | KLHDC4 | -53.7kb | Anti-sense | Chr. 20 - 74715381 - 74777248 on NCBI Build Mmul_10 | |
Downstream | solute carrier family 7 member 5 | SLC7A5 | 19.9kb | Anti-sense | Chr. 20 - 74850966 - 74891589 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
