Assay Details
Gene Symbol: MIR611 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032645.1 | 1 | 30 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | flap structure-specific endonuclease 1 | FEN1 | EGK_06132 | Anti-sense | Chr. 14 - 12377451 - 12382503 on NCBI Build Mmul_10 | |
Downstream | myelin regulatory factor | MYRF |
C11orf9, C14H11orf9 |
4.3kb | Anti-sense | Chr. 14 - 12386429 - 12423099 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
