Assay Details
Gene Symbol: MIR625 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032652.2 | 1 | 43 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | solute carrier family 24 member 1 | SLC24A1 | -94.4kb | Anti-sense | Chr. 7 - 42164548 - 42208542 on NCBI Build Mmul_10 | |
Downstream |
RAB11A, member RAS oncogene family |
RAB11A | EGK_17606 | 129.3kb | Anti-sense | Chr. 7 - 42432414 - 42453767 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
