Assay Details
Gene Symbol: MIR642 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032663.1 | 1 | 40 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | DAN domain BMP antagonist family member 5 | DAND5 | -10.1kb | Anti-sense | Chr. 19 - 12557835 - 12563371 on NCBI Build Mmul_10 | |
Downstream | nuclear factor I X | NFIX | 12.7kb | Anti-sense | Chr. 19 - 12586239 - 12689842 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
