Assay Details
Gene Symbol: MIR648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032666.2 | 1 | 45 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | proteasome inhibitor subunit 1 | PSMF1 | -49.6kb | Sense | Chr. 10 - 65386190 - 65433532 on NCBI Build Mmul_10 | |
Downstream | R-spondin 4 | RSPO4 | 66.0kb | Anti-sense | Chr. 10 - 65549225 - 65590871 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
