Assay Details
Gene Symbol: MIR888 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032698.1 | 1 | 37 | 68 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Rho GTPase activating protein 6 | ARHGAP6 | -63.2kb | Anti-sense | Chr. X - 10745388 - 11292172 on NCBI Build Mmul_10 | |
Downstream | MSL complex subunit 3 | MSL3 | MSL3L1 | 32.5kb | Sense | Chr. X - 11387966 - 11405035 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
