Assay Details
Gene Symbol: MIR942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032713.1 | 1 | 45 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif containing 45 | TRIM45 | EGK_01146 | -16.4kb | Anti-sense | Chr. 1 - 107141056 - 107151848 on NCBI Build Mmul_10 |
Downstream | CD101 molecule | CD101 | 57.6kb | Sense | Chr. 1 - 107226010 - 107264572 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
