Assay Details
Gene Symbol: MIR1235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032307.1 | 1 | 41 | 69 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | plectin | PLEC | PLEC1 | Sense | Chr. 8 - 144185448 - 144247047 on NCBI Build Mmul_10 | |
Downstream | microRNA mir-661 | MIR661 | mml-mir-661 | 17.3kb | Sense | Chr. 8 - 144215053 - 144215150 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
