Assay Details
Gene Symbol: MIR1239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032308.1 | 1 | 53 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
branched chain keto acid dehydrogenase E1, alpha polypeptide |
BCKDHA | Anti-sense | Chr. 19 - 40643382 - 40674275 on NCBI Build Mmul_10 | ||
Downstream | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 8 | B3GNT8 | EGK_10652 | 17.4kb | Sense | Chr. 19 - 40674631 - 40676953 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
