Assay Details
Gene Symbol: MIR1240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032309.1 | 1 | 46 | 57 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | autophagy related 9A | ATG9A | EGK_04816 | Sense | Chr. 12 - 106655612 - 106665661 on NCBI Build Mmul_10 | |
Downstream | ankyrin repeat and zinc finger peptidyl tRNA hydrolase 1 | ANKZF1 | 5.7kb | Anti-sense | Chr. 12 - 106665703 - 106673138 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
