Assay Details
Gene Symbol: MIR1241 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032310.1 | 1 | 45 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | endoplasmic reticulum to nucleus signaling 2 | ERN2 | Sense | Chr. 20 - 22034918 - 22058927 on NCBI Build Mmul_10 | ||
Downstream | calcineurin like EF-hand protein 2 | CHP2 | EGK_12607 | 56.1kb | Anti-sense | Chr. 20 - 22098670 - 22102957 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
