Assay Details
Gene Symbol: MIR510 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032318.1 | 1 | 54 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | midline 1 | MID1 | Anti-sense | Chr. X - 10025730 - 10416839 on NCBI Build Mmul_10 | ||
Downstream | holocytochrome c synthase | HCCS | 633.6kb | Sense | Chr. X - 10719443 - 10731226 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
