Assay Details
Gene Symbol: MIR133C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_032403.1 | 1 | 45 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | bromodomain containing 1 | BRD1 | -96.3kb | Anti-sense | Chr. 10 - 1650338 - 1702146 on NCBI Build Mmul_10 | |
Downstream | chromosome 10 C22orf34 homolog | C10H22orf34 | 57.2kb | Anti-sense | Chr. 10 - 1855698 - 1859490 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
