Assay Details
Gene Symbol: MIR6129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128322.1 | 1 | 55 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | chromosome 16 C17orf50 homolog | C16H17orf50 | EGK_08416 | -5.4kb | Anti-sense | Chr. 16 - 32107332 - 32111517 on NCBI Build Mmul_10 |
Downstream | TATA-box binding protein associated factor 15 | TAF15 | 37.9kb | Anti-sense | Chr. 16 - 32154897 - 32233544 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
