Assay Details
Gene Symbol: MIR7171 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128346.1 | 1 | 36 | 58 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
sodium leak channel, non-selective |
NALCN | -124.5kb | Sense | Chr. 17 - 82744571 - 83099029 on NCBI Build Mmul_10 | |
Downstream | fibroblast growth factor 14 | FGF14 | 158.0kb | Sense | Chr. 17 - 83381664 - 84057490 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
