Assay Details
Gene Symbol: MIR7184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_128367.1 | 1 | 33 | 55 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
anterior gradient 2, protein disulphide isomerase family member |
AGR2 | -3.5kb | Anti-sense | Chr. 3 - 98199805 - 98213070 on NCBI Build Mmul_10 | |
Downstream | tetraspanin 13 | TSPAN13 | EGK_13885 | 1.6kb | Sense | Chr. 3 - 98218298 - 98248497 on NCBI Build Mmul_10 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
