Assay Details
Gene Symbol: NPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194502.3 | NP_001181431.2 | 4 - 5 | 441 | 68 | |
GenBank mRNA | JU327025.1 | - | 4 - 5 | 418 | 68 | |
JV049568.1 | - | 4 - 5 | 422 | 68 | ||
JV565029.1 | - | 4 - 5 | 429 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
