Assay Details
Gene Symbol: INSL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204361.1 | NP_001191290.1 | 1 - 2 | 194 | 90 | |
GenBank mRNA | HM102325.1 | - | 1 - 2 | 194 | 90 |
Gene Symbol: LOC114675602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028842821.1 | XP_028698654.1 | 212 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
