Assay Details
Gene Symbol: LOC107000597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015149832.2 | XP_015005318.2 | 1648 | 101 | ||
GenBank mRNA | JU485864.1 | - | 1 | 223 | 101 |
Gene Symbol: LOC114670534 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028828622.1 | XP_028684455.1 | 370 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
