Assay Details
Gene Symbol: PRSS41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841589.1 | XP_028697422.1 | 346 | 67 | ||
GenBank mRNA | JU485932.1 | - | 1 - 2 | 129 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
