Assay Details
Gene Symbol: KLF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808151.3 | XP_002808197.2 | 335 | 96 | ||
GenBank mRNA | JU492293.1 | - | 1 | 121 | 96 | |
JV659829.1 | - | 1 | 114 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002808151.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
