Assay Details
Gene Symbol: SLC35E2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133065.2 | XP_014988551.2 | 1824 | 95 | ||
GenBank mRNA | JU336202.1 | - | 7 - 8 | 1226 | 95 | |
JU470982.1 | - | 6 - 7 | 961 | 95 | ||
JV044391.1 | - | 7 - 8 | 1037 | 95 | ||
JV564969.1 | - | 7 - 8 | 1197 | 95 | ||
JV627632.1 | - | 7 - 8 | 1250 | 95 | ||
JV667655.1 | - | 7 - 8 | 1291 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
