Assay Details
Gene Symbol: ZNF446 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125002.2 | XP_014980488.2 | 1489 | 109 | ||
XM_015125003.2 | XP_014980489.2 | 1114 | 109 | |||
XM_015125005.2 | XP_014980491.2 | 1520 | 109 | |||
XM_028839815.1 | XP_028695648.1 | 920 | 109 | |||
XM_028839816.1 | XP_028695649.1 | 933 | 109 | |||
GenBank mRNA | JU489459.1 | - | 5 - 6 | 836 | 109 | |
JV718477.1 | - | 6 - 7 | 896 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
