Assay Details
Gene Symbol: MRM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JU335253.1 | - | 1 - 2 | 319 | 79 | |
JU476223.1 | - | 1 - 2 | 327 | 79 | ||
JV047519.1 | - | 1 - 2 | 318 | 79 | ||
JV620980.1 | - | 1 - 2 | 340 | 79 | ||
JV636251.1 | - | 1 - 2 | 328 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
