Assay Details
Gene Symbol: CBARP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028838766.1 | XP_028694599.1 | 1130 | 92 | ||
XM_028838767.1 | XP_028694600.1 | 736 | 92 | |||
XM_028838768.1 | XP_028694601.1 | 767 | 92 | |||
GenBank mRNA | JU329207.1 | - | 4 - 5 | 427 | 92 | |
JV577254.1 | - | 5 - 6 | 522 | 92 | ||
JV679138.1 | - | 5 - 6 | 524 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
