Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151298.2 | XP_015006784.1 | 437 | 85 | ||
XM_015151299.2 | XP_015006785.1 | 437 | 85 | |||
XM_015151305.2 | XP_015006791.1 | 283 | 85 | |||
XM_015151306.2 | XP_015006792.1 | 302 | 85 | |||
XM_015151307.2 | XP_015006793.1 | 283 | 85 | |||
GenBank mRNA | JU486618.1 | - | 1 | 222 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015151298.2, XM_015151299.2, XM_015151305.2, XM_015151306.2, XM_015151307.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
