Assay Details
Gene Symbol: GPRC5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028841432.1 | XP_028697265.1 | 1357 | 66 | ||
XM_028841433.1 | XP_028697266.1 | 1698 | 66 | |||
XM_028841434.1 | XP_028697267.1 | 1238 | 66 | |||
GenBank mRNA | JV044395.1 | - | 3 - 4 | 1274 | 66 | |
JV653563.1 | - | 3 - 4 | 1186 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
