Assay Details
Gene Symbol: CIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808014.3 | XP_002808060.3 | 418 | 81 | ||
GenBank mRNA | JU496728.1 | - | 5 - 6 | 328 | 81 | |
JV727454.1 | - | 5 - 6 | 397 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
