Assay Details
Gene Symbol: BAG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002805223.3 | XP_002805269.1 | 334 | 100 | ||
GenBank mRNA | JU350898.1 | - | 1 - 2 | 82 | 100 | |
JV703900.1 | - | 1 - 2 | 204 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002805223.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
