Assay Details
Gene Symbol: BRI3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002798836.3 | XP_002798882.3 | 362 | 109 | ||
GenBank mRNA | JU474375.1 | - | 1 - 2 | 281 | 109 | |
JV642776.1 | - | 1 - 2 | 219 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
