Assay Details
Gene Symbol: IRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015111186.2 | XP_014966672.1 | 5034 | 91 | ||
GenBank mRNA | JV543865.1 | - | 83 | 91 | ||
JV650567.1 | - | 327 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
