Assay Details
Gene Symbol: POLR2J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015134548.2 | XP_014990034.1 | 481 | 84 | ||
GenBank mRNA | JU319779.1 | - | 3 - 4 | 432 | 84 | |
JV045614.1 | - | 3 - 4 | 423 | 84 | ||
JV602330.1 | - | 3 - 4 | 357 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
