Assay Details
Gene Symbol: SLC2A4RG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028828744.1 | XP_028684577.1 | 781 | 90 | ||
GenBank mRNA | JV656317.1 | - | 5 - 6 | 663 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
