Assay Details
Gene Symbol: ZNF366 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015140265.2 | XP_014995751.2 | 1890 | 97 | ||
GenBank mRNA | JU481568.1 | - | 3 - 4 | 597 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
