Assay Details
Gene Symbol: ALKAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028832030.1 | XP_028687863.1 | 357 | 129 | ||
XM_028832031.1 | XP_028687864.1 | 357 | 129 | |||
GenBank mRNA | JV648964.1 | - | 1 - 2 | 130 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() |
