Assay Details
Gene Symbol: PCDH11X | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015127772.2 | XP_014983258.1 | 3454 | 93 | ||
XM_028842285.1 | XP_028698118.1 | 3296 | 93 | |||
XM_028842286.1 | XP_028698119.1 | 3296 | 93 | |||
XM_028842287.1 | XP_028698120.1 | 3296 | 93 | |||
XM_028842288.1 | XP_028698121.1 | 3296 | 93 | |||
GenBank mRNA | JU334032.1 | - | 2 - 3 | 3081 | 93 | |
JU334033.1 | - | 2 - 3 | 3081 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
