Assay Details
Gene Symbol: REXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001088611.4 | XP_001088611.2 | 434 | 99 | ||
XM_001088838.4 | XP_001088838.2 | 521 | 99 | |||
XM_015125750.2 | XP_014981236.2 | 464 | 99 | |||
XM_015125751.2 | XP_014981237.2 | 439 | 99 | |||
GenBank mRNA | JV662616.1 | - | 3 - 4 | 360 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
