Assay Details
Gene Symbol: PIFO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148174.2 | XP_015003660.1 | 458 | 85 | ||
XM_015148180.2 | XP_015003666.1 | 621 | 85 | |||
GenBank mRNA | JU332916.1 | - | 3 - 4 | 293 | 85 | |
JU480277.1 | - | 2 - 3 | 198 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
