Assay Details
Gene Symbol: PGAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015133659.2 | XP_014989145.1 | 2534 | 82 | ||
GenBank mRNA | JU334989.1 | - | 2 - 3 | 656 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
