Assay Details
Gene Symbol: PLA2G2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015129834.2 | XP_014985320.1 | 392 | 115 | ||
XM_028838580.1 | XP_028694413.1 | 854 | 115 | |||
GenBank mRNA | JU470687.1 | - | 3 - 4 | 421 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
