Assay Details
Gene Symbol: ETV3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112523.2 | XP_014968009.1 | 278 | 68 | ||
XM_015112532.2 | XP_014968018.1 | 235 | 68 | |||
GenBank mRNA | JU333365.1 | - | 1 - 2 | 168 | 68 | |
JU474659.1 | - | 1 - 2 | 87 | 68 | ||
JV046434.1 | - | 1 - 2 | 285 | 68 | ||
JV645040.1 | - | 1 - 2 | 266 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015112523.2, XM_015112532.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
