Assay Details
Gene Symbol: LOC106995404 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015128651.2 | XP_014984137.1 | 857 | 100 | ||
GenBank mRNA | FJ527024.1 | - | 1 - 2 | 740 | 100 | |
JV726544.1 | - | 1 - 2 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
