Assay Details
Gene Symbol: CATSPERB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002808468.3 | XP_002808514.3 | 2086 | 129 | ||
XM_028851766.1 | XP_028707599.1 | 2065 | 129 | |||
XM_028851767.1 | XP_028707600.1 | 1937 | 129 |

More Information
Set Membership: |
![]() ![]() |
