Assay Details
Gene Symbol: CPSF4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028836742.1 | XP_028692575.1 | 2186 | 75 | ||
XM_028836743.1 | XP_028692576.1 | 2197 | 75 | |||
XM_028836744.1 | XP_028692577.1 | 2189 | 75 | |||
XM_028836745.1 | XP_028692578.1 | 458 | 75 | |||
XM_028836746.1 | XP_028692579.1 | 2165 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
